ENA946831491 Enamine

(2,2,5,5-tetramethyloxolan-3-yl)methanesulfonyl chloride

  • MDL number MFCD30668584
  • Cas Number : 2090313-43-0
  • Molecular Weight : 240.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-313674

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-313674
Availability details for
Quantity
  • Availability
  • Add to Cart