ENA946831497 Enamine

3-(2,2-dimethoxyethyl)-1-phenyl-5-(trifluoromethyl)-1H-pyrazole

  • MDL number MFCD30730438
  • Cas Number : 1482520-12-6
  • Molecular Weight : 300.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-314799

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-314799
Availability details for
Quantity
  • Availability
  • Add to Cart