ENA946831498 Enamine

N-(3-chloropropyl)-N-methylcarbamoyl chloride

  • MDL number MFCD30730440
  • Cas Number : 2095408-95-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -10
Physical Form liquid
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-315871

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-315871
Availability details for
Quantity
  • Availability
  • Add to Cart