ENA946831500 Enamine

tert-butyl 4-{4-oxo-1H,4H,5H-pyrazolo[3,4-d]pyrimidin-1-yl}piperidine-1-carboxylate

  • MDL number MFCD30730442
  • Cas Number : 2095410-97-0
  • Molecular Weight : 319.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-317260

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-317260
Availability details for
Quantity
  • Availability
  • Add to Cart