ENA946831502 Enamine

3-ethynyl-2-methyl-4,5,6,7-tetrahydro-2H-indazole

  • MDL number MFCD30695094
  • Cas Number : 2094471-58-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-320964

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-320964
Availability details for
Quantity
  • Availability
  • Add to Cart