ENA946831511 Enamine

4-methoxy-1,2-dimethylpyrrolidine-2-carboxylic acid hydrochloride

  • MDL number MFCD30730468
  • Cas Number : 2095409-12-2
  • Molecular Weight : 209.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-322259

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-322259
Availability details for
Quantity
  • Availability
  • Add to Cart