ENA946831513 Enamine

[(2R,4S)-4-methyl-2-(pyridin-3-yl)-3,4-dihydro-2H-1-benzopyran-4-yl]methanol

  • MDL number MFCD30730470
  • Cas Number : 2095396-08-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-322319

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-322319
Availability details for
Quantity
  • Availability
  • Add to Cart