ENA946831516 Enamine

2-[5-(2-methylpropyl)-1,3,4-oxadiazol-2-yl]-1H-indole

  • MDL number MFCD30730474
  • Cas Number : 2095410-17-4
  • Molecular Weight : 241.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-322401

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-322401
Availability details for
Quantity
  • Availability
  • Add to Cart