ENA946831517 Enamine

5-hydroxy-1-methyl-6-propyl-1H,2H,3H,4H-pyrido[2,3-d]pyrimidine-2,4-dione

  • MDL number MFCD30730475
  • Cas Number : 2095409-77-9
  • Molecular Weight : 235.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-322412

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-322412
Availability details for
Quantity
  • Availability
  • Add to Cart