ENA946831518 Enamine

6-[(3-methyl-1,2,4-oxadiazol-5-yl)methoxy]pyridine-3-carboxylic acid

  • MDL number MFCD30730476
  • Cas Number : 2095409-91-7
  • Molecular Weight : 235.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-322430

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-322430
Availability details for
Quantity
  • Availability
  • Add to Cart