ENA946831521 Enamine

6-fluoro-8-{[(3-methylpyridin-2-yl)oxy]methyl}quinoline

  • MDL number MFCD30730479
  • Cas Number : 2095410-77-6
  • Molecular Weight : 268.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-322440

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-322440
Availability details for
Quantity
  • Availability
  • Add to Cart