ENA946831523 Enamine

4-[(2-bromoethyl)sulfanyl]pyridine hydrobromide

  • MDL number MFCD30730481
  • Cas Number : 2095409-94-0
  • Molecular Weight : 299.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-322486

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-322486
Availability details for
Quantity
  • Availability
  • Add to Cart