ENA946831525 Enamine

3-[(2-bromophenyl)methyl]azetidine; trifluoroacetic acid

  • MDL number MFCD30730483
  • Cas Number : 2095408-96-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data CF3COOH
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-322511

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-322511
Availability details for
Quantity
  • Availability
  • Add to Cart