ENA946831530 Enamine

(1s,3s)-1-{[(tert-butoxy)carbonyl]amino}-3-hydroxycyclobutane-1-carboxylic acid

  • MDL number MFCD30730491
  • Cas Number : 2095396-14-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-323207

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-323207
Availability details for
Quantity
  • Availability
  • Add to Cart