ENA946831535 Enamine

(1s,3s)-3-hydroxy-1-phenylcyclobutane-1-carboxamide

  • MDL number MFCD30730495
  • Cas Number : 2091554-91-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-332473

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-332473
Availability details for
Quantity
  • Availability
  • Add to Cart