ENA946831542 Enamine

5-[(1E)-3-(tert-butoxy)-3-oxoprop-1-en-1-yl]furan-3-carboxylic acid

  • MDL number MFCD30730506
  • Cas Number : 2094962-39-5
  • Molecular Weight : 238.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-342557

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-342557
Availability details for
Quantity
  • Availability
  • Add to Cart