ENA946831544 Enamine

1-methanesulfonyl-2,5-dimethylpiperazine hydrochloride

  • MDL number MFCD30730508
  • Cas Number : 2095409-96-2
  • Molecular Weight : 228.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-343502

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-343502
Availability details for
Quantity
  • Availability
  • Add to Cart