ENA946831549 Enamine

3-[4-(trifluoromethyl)phenyl]propane-1-sulfonyl chloride

  • MDL number MFCD30730515
  • Cas Number : 2095409-80-4
  • Molecular Weight : 286.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-343791

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-343791
Availability details for
Quantity
  • Availability
  • Add to Cart