ENA946831557 Enamine

4-bromo-5-iodothiophene-3-carboxylic acid

  • MDL number MFCD30730523
  • Cas Number : 57233-96-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-345171

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-345171
Availability details for
Quantity
  • Availability
  • Add to Cart