ENA946831560 Enamine

1H,3H,4H-pyrrolo[2,1-c][1,4]oxazine-1,3-dione

  • MDL number MFCD30730527
  • Cas Number : 2095409-32-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-345825

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-345825
Availability details for
Quantity
  • Availability
  • Add to Cart