ENA946831561 Enamine

1,3-dichloro-6,7-difluoroisoquinoline

  • MDL number MFCD30730528
  • Cas Number : 2092310-58-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-345979

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-345979
Availability details for
Quantity
  • Availability
  • Add to Cart