ENA946831564 Enamine

4-(1,1,2,2,3,3,3-heptafluoropropoxy)-3-(propan-2-yl)benzene-1-sulfonyl chloride

  • MDL number MFCD30730532
  • Cas Number : 2095409-41-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-352271

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-352271
Availability details for
Quantity
  • Availability
  • Add to Cart