ENA946831570 Enamine

1-(trifluoromethyl)-5H,6H,7H,8H-imidazo[1,5-a]pyrazin-8-one

  • MDL number MFCD30730544
  • Cas Number : 2094627-94-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-365135

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-365135
Availability details for
Quantity
  • Availability
  • Add to Cart