ENA946831571 Enamine

tert-butyl N-[2-(4-ethyl-5-sulfanyl-4H-1,2,4-triazol-3-yl)ethyl]carbamate

  • MDL number MFCD30730546
  • Cas Number : 2095410-03-8
  • Molecular Weight : 272.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-365169

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-365169
Availability details for
Quantity
  • Availability
  • Add to Cart