ENA946831577 Enamine

2-[(tert-butyldimethylsilyl)oxy]-2-cyclopentylacetaldehyde

  • MDL number MFCD30730552
  • Cas Number : 2095409-48-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-365478

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-365478
Availability details for
Quantity
  • Availability
  • Add to Cart