ENA946831587 Enamine

octahydro-2lambda6-thieno[3,4-c]pyridine-2,2-dione hydrochloride

  • MDL number MFCD30730565
  • Cas Number : 2094843-87-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-365993

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-365993
Availability details for
Quantity
  • Availability
  • Add to Cart