ENA946831590 Enamine

3-(chloromethyl)-1-(trimethylsilyl)hept-1-yn-3-ol

  • MDL number MFCD30730568
  • Cas Number : 2095410-47-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-366054

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-366054
Availability details for
Quantity
  • Availability
  • Add to Cart