ENA946831591 Enamine

tert-butyl 1-(bromomethyl)-2-azabicyclo[2.1.1]hexane-2-carboxylate

  • MDL number MFCD30730569
  • Cas Number : 2002785-62-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -10
Physical Form oil
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-366068

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-366068
Availability details for
Quantity
  • Availability
  • Add to Cart