ENA946831595 Enamine

4-ethoxy-N-methylaniline hydrochloride

  • MDL number MFCD30730574
  • Cas Number : 15206-42-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-366185

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-366185
Availability details for
Quantity
  • Availability
  • Add to Cart