ENA946831599 Enamine

methyl 4-aminoimidazo[2,1-c][1,2,4]triazine-3-carboxylate

  • MDL number MFCD30730577
  • Cas Number : 2090997-95-6
  • Molecular Weight : 193.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-366206

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-366206
Availability details for
Quantity
  • Availability
  • Add to Cart