ENA946831605 Enamine

3-{1-[(1-methyl-1H-imidazol-2-yl)methyl]-4,5,6,7-tetrahydro-1H-1,3-benzodiazol-2-yl}propan-1-amine dihydrochloride

  • MDL number MFCD30730583
  • Cas Number : 2094623-89-7
  • Molecular Weight : 346.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-366264

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-366264
Availability details for
Quantity
  • Availability
  • Add to Cart