ENA946831606 Enamine

2-(iodomethyl)-hexahydro-2H-cyclopenta[b]furan

  • MDL number MFCD30730585
  • Cas Number : 123121-05-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-366293

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-366293
Availability details for
Quantity
  • Availability
  • Add to Cart