ENA946831613 Enamine

methyl 2-(1,3-dioxo-2,3-dihydro-1H-isoindol-2-yl)-5-hydroxybenzoate

  • MDL number MFCD30730595
  • Cas Number : 146041-93-2
  • Molecular Weight : 297.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-366343

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-366343
Availability details for
Quantity
  • Availability
  • Add to Cart