ENA946831616 Enamine

2-{1H-pyrazolo[3,4-b]pyridin-1-yl}acetic acid hydrochloride

  • MDL number MFCD30730601
  • Cas Number : 2093856-68-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-366380

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-366380
Availability details for
Quantity
  • Availability
  • Add to Cart