ENA946831623 Enamine

{7-oxabicyclo[2.2.1]heptan-1-yl}methanesulfonyl chloride

  • MDL number MFCD30730611
  • Cas Number : 2091597-91-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-366444

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-366444
Availability details for
Quantity
  • Availability
  • Add to Cart