ENA946831625 Enamine

2-[(1-methyl-1H-pyrazol-4-yl)formamido]propanoic acid hydrochloride

  • MDL number MFCD30730614

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-366487

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-366487
Availability details for
Quantity
  • Availability
  • Add to Cart