ENA946831634 Enamine

2-(4,5-dibromothiophen-2-yl)ethan-1-amine hydrochloride

  • MDL number MFCD30730628
  • Cas Number : 2095410-21-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-366592

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-366592
Availability details for
Quantity
  • Availability
  • Add to Cart