ENA946831645 Enamine

(6,6-dimethyloxan-2-yl)methanesulfonamide

  • MDL number MFCD30730639
  • Cas Number : 2095410-29-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-366697

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-366697
Availability details for
Quantity
  • Availability
  • Add to Cart