ENA946831646 Enamine

2-chloro-5-(dibromomethyl)-1,3-thiazole

  • MDL number MFCD30730641
  • Cas Number : 2092310-61-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-366708

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-366708
Availability details for
Quantity
  • Availability
  • Add to Cart