ENA946831657 Enamine

2,3-dichloro-5-{4-[(dimethylamino)methyl]piperidine-1-carbonyl}aniline dihydrochloride

  • MDL number MFCD30730658
  • Cas Number : 2095409-13-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-366820

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-366820
Availability details for
Quantity
  • Availability
  • Add to Cart