ENA946831661 Enamine

7-amino-3,4-dihydro-2H-1lambda6,4-benzothiazine-1,1,3-trione hydrochloride

  • MDL number MFCD30730662
  • Cas Number : 2095409-15-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-366840

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-366840
Availability details for
Quantity
  • Availability
  • Add to Cart