ENA946831662 Enamine

2-(3-methoxyazetidin-3-yl)-1-methyl-1H-imidazole dihydrochloride

  • MDL number MFCD30730664
  • Cas Number : 2094615-71-9
  • Molecular Weight : 240.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-366843

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-366843
Availability details for
Quantity
  • Availability
  • Add to Cart