ENA946831672 Enamine

1-(2-ethyl-5-methyl-2H-1,2,3-triazol-4-yl)ethan-1-one

  • MDL number MFCD30730402
  • Cas Number : 1934906-01-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-226491

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-226491
Availability details for
Quantity
  • Availability
  • Add to Cart