ENA946831678 Enamine

5-oxaspiro[3.5]nonane-8-carbonitrile

  • MDL number MFCD23792803
  • Cas Number : 1503527-20-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-243980

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-243980
Availability details for
Quantity
  • Availability
  • Add to Cart