ENA946831680 Enamine

2-(azepan-4-yl)acetamide hydrochloride

  • MDL number MFCD29970418
  • Cas Number : 1862903-99-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-244172

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-244172
Availability details for
Quantity
  • Availability
  • Add to Cart