ENA946831684 Enamine

4-methoxy-3,3-dimethylbutanenitrile

  • MDL number MFCD27930058
  • Cas Number : 854866-84-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-244931

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-244931
Availability details for
Quantity
  • Availability
  • Add to Cart