ENA946831693 Enamine

decahydro-1,7-naphthyridin-8-one

  • MDL number MFCD28970431
  • Cas Number : 1922714-98-4
  • Molecular Weight : 154.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-249653

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-249653
Availability details for
Quantity
  • Availability
  • Add to Cart