ENA946831702 Enamine

[4-(1,3-dioxolan-2-yl)-1,3-thiazol-2-yl]methanol

  • MDL number MFCD30730407
  • Cas Number : 1934407-64-3
  • Molecular Weight : 187.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-252069

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-252069
Availability details for
Quantity
  • Availability
  • Add to Cart