ENA946831718 Enamine

1-(1-methyl-1H-1,2,3-triazol-4-yl)ethan-1-ol

  • MDL number MFCD24109419
  • Cas Number : 1022093-96-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-261370

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-261370
Availability details for
Quantity
  • Availability
  • Add to Cart