ENA946831741 Enamine

3-ethyl-1-methyl-1H-pyrazole-4-sulfonyl chloride

  • MDL number MFCD08457190
  • Cas Number : 1314922-70-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-272547

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-272547
Availability details for
Quantity
  • Availability
  • Add to Cart